Tlačiť
Watch glasses
Dodávateľ: KARTELL
452 454 455 456
216-1134EA 5.05 EUR
216-1134 216-1136 216-1138 216-1140
Watch glasses
Skla hodinová
PP, transparent.
  • Excellent chemical resistance
  • Heat resistant up to +120 °C
  • Autoclavable
Stable, with extra wide foot.