Tlačiť
Sample bottles with screw cap
Dodávateľ: MUELLER KREMPEL
14.020.02 14.020.01 14.020.03 14.020.04 14.020.05
215-8341EA 417.17 EUR
215-8341 215-8342 215-8343 215-8344 215-8345
Sample bottles with screw cap
Vialky
Vials made from AR glass, clear with black screw cap. Ideal for taking, storing and transporting samples.
  • Thin-walled, cylindrical
  • Tightly sealing screw cap
  • Designed for use in applications that do not require resistance to temperature