Tlačiť
Sample vials, trident vials, squat or tall
Dodávateľ: MURRAY S
T103/V4T004/C T103/V2T004/C T105/V2T005/C T103/V1T004/C T105/V1T005/C T105/V3T005/C T103/V3T004/C
212-7069EA 182.11 EUR
212-7069 212-7066 212-7072 212-7064 212-7071 212-7073 212-7068
Sample vials, trident vials, squat or tall
Vialky
Vials made from neutral glass with PE plug stoppers. Suitable for a wide variety of uses in the laboratory, production, more.
  • Robust and economical, will withstand moderate temperature change
  • Wide opening for easy filling and emptying
  • Plug stopper for secure closure
Balenie: For production related reasons, the number of vials per box may vary.