Tlačiť
Vials with screw thread, Nalgene®
Dodávateľ: Thermo Fisher Scientific
342032-0020 322032-0005 322032-0010 322032-0020 342032-0005 342032-0010 342033-0010
215-1499EA 691.54 EUR
215-1499 215-1486 215-1487 215-1488 215-1497 215-1498 215-1505
Vials with screw thread, Nalgene®
Vialky
These vials are ideal for diagnostic and biological sera, pharmaceuticals for veterinary and ophthalmologic usage as well as many other applications. They are made from clear or coloured brown PETG.
  • Available clear or coloured brown, sterile or non-sterile
  • A wide range of different screw caps is optionally available
  • Manufactured and packed in workrooms equipped with HEPA filters
Closures for vials: HDPE, coloured. For safe and tight closure of narrow-neck vials. Available in non-sterile or sterile versions. Non sterile caps are loose packed, sterile caps are loose packed in a box with a three-section insert.
For safe and tight closure of narrow-neck vials.