Tlačiť
Colour coders for cryovials, CryoColorCode, Nunc™
Dodávateľ: Thermo Fisher Scientific
354755 354968 355077 375884 375922 375868 375930 354879 355018 355158 375906
479-2481EA 16.93 EUR
479-2481 479-6000 479-2482 479-2485 479-2487 479-2484 479-2488 479-6859 479-6860 479-6861 479-2486
Colour coders for cryovials, CryoColorCode, Nunc™
Vložky čepice
Colour coded inserts for the cap with inscription area.
Notch to remove tube or CryoColorCodes.