Tlačiť
Cryo.S™ tubes, 1 and 2 ml
Dodávateľ: Greiner Bio-One
122277_500 122278_500 122279_500 122280_500 123263_500 123277_500 123278_500 123279_500 123280_500 121263_500 121277_500 121278_500 121279_500 121280_500 126263_500 126277_500 126278_500 126279_500 126280_500 122263_500 121261_500 122261_500 126261_500 123261_500
391-3461EA 265.54 EUR
391-3461 391-3462 391-3463 391-3464 479-4110 479-4111 479-4112 479-4113 479-4114 479-4120 479-4121 479-4122 479-4123 479-4124 479-4140 479-4141 479-4142 479-4143 479-4144 479-0086 479-0099 479-0190 479-0191 GREI123261_500
Cryo.S™ tubes, 1 and 2 ml
Vialky
For storage in freezer or in the liquid nitrogen vapour phase. Thermal resistance of −196 to +121 °C.
  • Certified as non cytotoxic and free from endotoxins, DNase, RNase and human DNA
  • New graduation and marking field
  • Compatible with air transport
  • 1D and 2D bar codes on request
In PP, with a coloured cap, sterile (SAL 10⁻³ ).
* without writing area
** with Datamatrix code