Tlačiť
Capillary tubing
Dodávateľ: VICI JOUR RESEARCH
JR-T-6007-M3 JR-T-6008-M3 JR-T-6000-M3 JR-T-6001-M3 JR-T-6002-M3 JR-T-6003-M3 JR-T-60031-M3 JR-T-60032-M3 JR-T-5999-M3 JR-T-6009-M3 JR-T-6010-M3 JR-T-6011-M3 JR-T-6007-M10 JR-T-6008-M10 JR-T-60032-M10 JR-T-60031-M10 JR-T-6002-M10 JR-T-6001-M10 JR-T-6000-M10 JR-T-5999-M10 JR-T-6003-M10 JR-T-6011-M10 JR-T-6010-M10 JR-T-6009-M10 JR-T-6003-M25 JR-T-6003-M5 JR-T-6000-M5 JR-T-6001-M25 JR-T-6001-M5 JR-T-6002-M25 JR-T-6002-M5 JR-T-60031-M25 JR-T-60031-M5 JR-T-60032-M25 JR-T-60032-M5 JR-T-5802-M3 JR-T-5802-M25 JR-T-5802-M5 JR-T-5803-M10 JR-T-5803-M25 JR-T-5803-M3 JR-T-5803-M5 JR-T-5804-M10 JR-T-5804-M25 JR-T-5804-M3 JR-T-5804-M5 JR-T-5802-M10 JR-T-5999-M25 JR-T-5999-M5 JR-T-6000-M25
554-1086EA 62.17 EUR
554-1086 554-1087 554-1081 554-1082 554-1083 554-1084 554-1085 546-1236 554-1080 554-1088 554-1089 554-1090 JOURJR-T-6007-M10 JOURJR-T-6008-M10 JOURJR-T-60032-M10 JOURJR-T-60031-M10 JOURJR-T-6002-M10 JOURJR-T-6001-M10 JOURJR-T-6000-M10 JOURJR-T-5999-M10 JOURJR-T-6003-M10 JOURJR-T-6011-M10 JOURJR-T-6010-M10 JOURJR-T-6009-M10 JOURJR-T-6003-M25 JOURJR-T-6003-M5 JOURJR-T-6000-M5 JOURJR-T-6001-M25 JOURJR-T-6001-M5 JOURJR-T-6002-M25 JOURJR-T-6002-M5 JOURJR-T-60031-M25 JOURJR-T-60031-M5 JOURJR-T-60032-M25 JOURJR-T-60032-M5 JOURJR-T-5802-M3 JOURJR-T-5802-M25 JOURJR-T-5802-M5 JOURJR-T-5803-M10 JOURJR-T-5803-M25 JOURJR-T-5803-M3 JOURJR-T-5803-M5 JOURJR-T-5804-M10 JOURJR-T-5804-M25 JOURJR-T-5804-M3 JOURJR-T-5804-M5 JOURJR-T-5802-M10 JOURJR-T-5999-M25 JOURJR-T-5999-M5 JOURJR-T-6000-M25
Capillary tubing
Hadičky Chromatography Tubing
PEEK.
  • Withstand continuous pressure without swelling
  • Non-permeable to organic solvents
  • Inert and biocompatible
  • Colour coded for each diameter
Unaffected by high strength buffers, halide salts or aggressive mobile phases that would degrade stainless steel.
Pozor: Attention: Dichloromethane, THF and DMSO may cause swelling in PEEK. Concentrated nitric acid and sulphuric acid will attack PEEK.