Tlačiť
Capillary tubing
Dodávateľ: VICI JOUR RESEARCH
JR-T-4011-M3 JR-T-4036-M3 JR-T-4037-M3 JR-T-6807-M3 JR-T-4183-M3 JR-T-6800-M3 JR-T-6805-M3 JR-T-6807-M25 JR-T-6807-M10 JR-T-6805-M25 JR-T-6810 JR-T-6805-M10 JR-T-6800-M25 JR-T-4011-M25 JR-T-6800-M10 JR-T-4183-M25 JR-T-4183-M10 JR-T-4011-M10 JR-T-4037-M25 JR-T-4037-M10 JR-T-4036-M25 JR-T-4036-M10 JR-T-4036-M5 JR-T-4011-M5 JR-T-4037-M5 JR-T-4183-M5 JR-T-6800-M5 JR-T-6805-M5 JR-T-6807-M5
554-1060EA 21.5 EUR
554-1060 554-1061 554-1062 554-1100 554-1065 554-1098 554-1099 JOURJR-T-6807-M25 JOURJR-T-6807-M10 JOURJR-T-6805-M25 JOURJR-T-6810 JOURJR-T-6805-M10 JOURJR-T-6800-M25 JOURJR-T-4011-M25 JOURJR-T-6800-M10 JOURJR-T-4183-M25 JOURJR-T-4183-M10 JOURJR-T-4011-M10 JOURJR-T-4037-M25 JOURJR-T-4037-M10 JOURJR-T-4036-M25 JOURJR-T-4036-M10 JOURJR-T-4036-M5 JOURJR-T-4011-M5 JOURJR-T-4037-M5 JOURJR-T-4183-M5 JOURJR-T-6800-M5 JOURJR-T-6805-M5 JOURJR-T-6807-M5
Capillary tubing
Hadičky Chromatography Tubing
PTFE.
  • Chemically inert and biocompatible
  • Suitable for low and mid pressure applications