Tlačiť
Dialysis sacks, ready to use, Spectra/Por®
Dodávateľ: SPECTRUM CHROMATOGRAPHY
132666 132681 132726
734-0668EA 561.59 EUR
734-0668 734-0674 734-0688
Dialysis sacks, ready to use, Spectra/Por®
Hadičky Hadičky dializační
Regenerated cellulose.
The dialysis tubing is sealed at one end with a closure and open at the other. A funnel is attached to the top of the sack for easily filling.
Supplied wet in 0,1% sodium azide. Should be rinsed prior to use.
Length: 600 mm