Tlačiť
Tubing, PVC
Dodávateľ: DEUTSCH NEUMANN
3503545 3504050 3500509 3500507 3501016 3500118 3500204A 3500304B 3500305A 3500306B 3500406 3500407B 3500508A 3500608B 3500609 3500812B 3500912B 3501014B 3501016B 3501216 3501317 3501519A 3502026 3503039 3501521 3501013 3501215 3501620 3500408 3501319 3502534 3500203 3500405 3501218 3501927 3502228
228-1725EA 45.73 EUR
228-1725 228-1726 228-1751 228-1752 228-3771 228-3830 228-3831 228-3832 228-3833 228-3834 228-3835 228-3836 228-3837 228-3838 228-3839 228-3845 228-3846 228-3848 228-3849 228-3850 228-3851 228-3852 228-3854 228-3855 228-3857 228-1560 228-1561 228-1564 228-1559 228-1563 228-1568 228-1557 228-1558 228-1562 228-1566 228-1567
Tubing, PVC
Hadičky Přenosová hadička
PVC, transparent.
  • High flexibility, good chemical resistance
  • Smooth both inside and out, excellent resistance to abrasion
  • Good value, large size rolls
Universal tubing for a wide range of applications. The preferred option where the flow of liquids has to be observed. Not suitable for operation under vacuum or pressure.
Temperature resistance: −20 to +60 °C
Hardness (Shore durometer A): 77