Tlačiť
Metal tubing with small flanges
Dodávateľ: VACUUBRAND
20673305 20673306 20673307 20673308 20673315 20673316 20673317 20673318 20673325 20673326 20673327 20673328 20673335 20673336 20673337 20673338
185-2511EA 127.78 EUR
185-2511 185-2521 185-2531 185-2541 185-2512 185-2522 185-2532 185-2542 185-2513 185-2523 185-2533 185-2543 185-2514 185-2524 185-2534 185-2544
Metal tubing with small flanges
Hadičky Hadičky vakuové/tlakové
Stainless steel.
  • Meets strict gas discharge and cleanliness requirements
  • Simple, easy assembly and disassembly
  • Secure, vacuum tight connection in conjunction with optional centering and clamping rings
Highly flexible corrugated metal tubes with small flanges welded on both ends.