Tlačiť
Drying tubes with standard ground joints, angled
Dodávateľ: LENZ LABORGLAS
05337001 05337002 05337003 05337004
201-5311EA 23.87 EUR
201-5311 201-1166 201-1167 201-5312
Drying tubes with standard ground joints, angled
Zkumavky Trubice sušicí
Clear DURAN® borosilicate 3.3 glass. Angled at 75° with ground joint cone.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Suitable for use with a desiccant such as calcium chloride.