Tlačiť
Test tubes with standard ground stoppers
Dodávateľ: LENZ LABORGLAS
05700107 05700114 05700130 05700110 05700126 05700134 05700138 05700142
212-1622EA 6.18 EUR
212-1622 212-1624 212-1626 212-0650 212-0651 212-0652 212-0653 212-0654
Test tubes with standard ground stoppers
Zkumavky Zkumavky na vzorky
DURAN® borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Standard ground joint and HDPE stoppers, non graduated.