Tlačiť
Test tubes with Quickfit® ground sockets
Dodávateľ: DWK LIFE SCIENCES
MF24/12/4 MF24/1 MF24/1/5 MF24/1/6 MF24/3 MF24/0/4 MF24/2 MF24/2/6 MF24/3/8 MF24/5
201-8623EA 93.06 EUR
201-8623 201-8620 201-8621 201-8622 201-8626 201-8619 201-8624 201-8625 201-8627 201-8628
Test tubes with Quickfit® ground sockets
Zkumavky Zkumavky na vzorky
Pyrex® borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Standard ground sockets.
Objednávacie informácie: Stoppers have to be ordered separately.