Tlačiť
Test tubes
Dodávateľ: KARTELL
88301 88307 88308 88310 88108 88306 88302 88320
212-6110EA 267.95 EUR
212-6110 212-0220 212-0221 212-6117 212-0682 212-0684 212-0683 KART88320
Test tubes
Zkumavky Zkumavky na vzorky
PS, transparent or PP, translucent.
  • PS: Good thermal and chemical resistance
  • PP: Excellent thermal and chemical resistance, autoclavable
  • Robust, economical
Universal tubes for a wide range of applications.