Tlačiť
Test tubes
Dodávateľ: Greiner Bio-One
160101_1600 169101_1500 187201_1500 187101_1500 160201_1600 115101_2000 136101_1400 112101_3600 112201_3600 115201_2000
212-3150EA 114.16 EUR
212-3150 212-3186 391-3469 212-0363 212-3110 212-3140 212-3146 212-3165 212-0361 GREI115201_2000
Test tubes
Zkumavky Zkumavky na vzorky
PS, clear or PP, transparent.
  • PP: good temperature resistance, good chemical resistance
  • PS: highly transparent, well suited to visual measurements
  • Matching push-in stoppers are optionally available
  • Can also be used as centrifuge tubes
Universal tubes for a wide range of laboratory and production applications.