Tlačiť
Test tubes without rim, heavy wall
Dodávateľ: DWK LIFE SCIENCES
1622/02M 1622/04M 1622/06M 1622/09M 1622/20M 1622/22M 1622/14M 1622/15M 1622/10M 1622/12M 1622/16M
212-7409EA 68.59 EUR
212-7409 212-7410 212-7411 212-7412 212-7416 212-7417 212-7555 212-7556 212-7413 212-7414 212-7415
Test tubes without rim, heavy wall
Zkumavky Zkumavky na vzorky
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
High quality test tubes designed and constructed for optimum performance. Medium walled, robust.
ISO 4142
Certifikácie: ISO 4142