Tlačiť
Test tubes with rim, medium wall
Dodávateľ: DWK LIFE SCIENCES
1620/15M 1620/02M 1620/04M 1620/06M 1620/10M 1620/12M 1620/16M 1620/20M 1620/22M 1620/09M
212-0631EA 141.6 EUR
212-0631 212-7402 212-7403 212-0446 212-7405 212-7406 212-7407 212-7408 212-0447 212-7404
Test tubes with rim, medium wall
Zkumavky Zkumavky na vzorky
PYREX®, borosilicate glass.
  • Superior performance in comparison to neutral borosilicate and soda-lime glass
  • Excellent resistance to chemicals and high temperature
High quality test tubes designed and constructed for optimum performance.
Certifikácie: ISO 4142