Tlačiť
Culture tubes, disposable, with screw cap, DURAN®
Dodávateľ: DWK LIFE SCIENCES
231751159 231751459 231752155 231752352
391-0140EA 69.8 EUR
391-0140 391-0145 391-0155 212-0237
Culture tubes, disposable, with screw cap, DURAN®
Zkumavky Zkumavky kultivační
Soda-lime glass with DIN thread and PP screw cap.
  • Economical disposable culture tubes
  • Food grade TPE gasket
For use in applications without high thermal loads.