Tlačiť
Culture tubes with screw cap, DURAN®
Dodávateľ: DWK LIFE SCIENCES
261352151 261352357 261351155 261351258 261352254 261352451
391-0171EA 360.21 EUR
391-0171 391-0172 391-4022 212-0375 212-0377 212-0379
Culture tubes with screw cap, DURAN®
Zkumavky Zkumavky kultivační
DURAN®, borosilicate glass 3.3, clear, with DIN thread and red PBT screw caps with PTFE coated gaskets.
  • Excellent chemical resistance
  • High thermal resistance, sterilisation possible up to 1 bar overpressure at 121 °C
  • Minimal thermal expansion giving relatively high resistance to temperature changes