Tlačiť
Microcentrifuge tubes, Micrewtube®
Dodávateľ: SIMPORT SCIENTIFIC
T336-2 T336-2S T336-4 T336-4S T336-5 T336-5S T336-6 T336-6S T336-7 T336-7S T336-2SPR T336-4SPR T336-5SPR T336-6SPR T336-7SPR
720-0522EA 209.39 EUR
720-0522 720-0523 720-0525 720-0526 720-0528 720-0529 720-0531 720-0532 212-9585 212-9586 720-0524 720-0527 720-0530 212-9584 525-0386
Microcentrifuge tubes, Micrewtube®
Zkumavky Mikrozkumavky centrifugační
PP, with lip seal screw cap and attachment loop.
  • Flexible sealing lip inside the cap ensures a positive leakproof seal under even the most adverse conditions
  • Sterile tubes are gamma irradiated and available either plain or with printed graduations and white marking area for sample identification
  • Certified RNase-, DNase-, pyrogen- and DNA-free
Ideal for freezer storage, boiling application, centrifugation etc. and will fit most standard microcentrifuge rotors. Caps are supplied with attachment loops in order to prevent contamination and mix-up.
Max. RCF: Non skirted tubes can be centrifuged up to 20000×g.
Temperature range: –90 to +100 °C
Balenie: Sterile tubes are packaged in tamper-proof resealable bags.