Tlačiť
Microcentrifuge tubes, 0,6 ml and 1,5 ml, Cliklok™
Dodávateľ: SIMPORT SCIENTIFIC
T330-8N T330-7G T330-7B T330-7N T330-7Y T330-6N T330-7AM
720-0506EA 30.45 EUR
720-0506 720-1010 720-1009 720-1011 720-1012 SIMPT330-6N SIMPT330-7AM
Microcentrifuge tubes, 0,6 ml and 1,5 ml, Cliklok™
Zkumavky Mikrozkumavky centrifugační
PP, graduated, with flat cap Cliklok™.
  • Can withstand ultra high centrifugation to 20000×g
  • Flat caps can be easily labelled
  • Certified RNase, DNase, pyrogen and DNA-free
  • Cliklok™ sealing caps (lock in two stages) reduce popping where freezing, boiling, storing or shipping is required
Balenie: Microtubes packed in resealable bags.