Tlačiť
Melting point capillaries
Dodávateľ: VITREX MEDICAL
224513 213413 216513 213613 213713 213813 213913 210713 210813 214313 211313 211413 211513 219513 219913 211213 211713 216613 210413 210913 211613 211113 213513 214513 217913 210513 218613 210113 210613 210213 210313 211013
MODU224513EA 30.85 EUR
MODU224513 621-1806 621-1812 621-1807 621-1808 621-1809 621-1810 621-1801 621-1802 621-1811 621-1803 621-1804 621-1805 MODU219513 MODU219913 MODU211213 MODU211713 MODU216613 MODU210413 MODU210913 MODU211613 MODU211113 MODU213513 MODU214513 MODU217913 MODU210513 MODU218613 MODU210113 MODU210613 MODU210213 MODU210313 MODU211013
Melting point capillaries
Zkumavky Kapiláry pro určení bodu tání
Borosilicate glass tubes. Uniform and cut at right angles.
Capillary tubes and melting point tubes are quality products designed for the handling and transport of small volumes of liquid, melting point determinations and many other purposes.
Balenie: Vials of 100 pieces or bulk packed in boxes.