Tlačiť
Sealing caps, Nalgene®
Dodávateľ: Thermo Fisher Scientific
71-3132-0058 71-3132-0063 DS3132-0020 DS3132-0024 DS3132-0058 DS3132-0063
NALG71-3132-0058EA 74.21 EUR
NALG71-3132-0058 NALG71-3132-0063 525-2923 525-2924 525-2925 525-2926
Sealing caps, Nalgene®
Uzávěry Víčka na zkumavky
PP, autoclavable.
  • Silicone gasket
For use at recommended maximum speeds 10000×g without leakage, even when full.