Tlačiť
Centrifuge tubes, Nalgene®
Dodávateľ: Thermo Fisher Scientific
3110-0120 3110-0150 3110-0160 3110-0380 3110-0500 3110-1000 3110-9500
525-2853EA 64.98 EUR
525-2853 525-2854 525-2855 525-2857 525-2858 525-2860 525-2859
Centrifuge tubes, Nalgene®
Zkumavky Zkumavky centrifugační
PPCO, round bottom and rimless, autoclavable.
  • Translucent, excellent chemical resistance
  • Withstand forces up to 50000×g
  • More suitable for low temperatures than PP
Objednávacie informácie: 525-2859 and 525-2860 are lipped.