Tlačiť
Cell culture tubes, Nunclon™Δ
Dodávateľ: Thermo Fisher Scientific
145470 146183 156758
734-2036EA 588.85 EUR
734-2036 734-2037 734-2068
Cell culture tubes, Nunclon™Δ
Zkumavky Zkumavky kultivační
PS with PE caps, sterile.
  • Flat side allows easy microscopy - A coverslip can be used in the tube
  • Medium retained in the flat-sided tube in the horizontal position
  • Excellent optical quality
  • Certified surface treatment for optimal cell attachment and growth
Round bottom tubes with screw or push-on cap for standard cell culture, or flat-sided tubes for the culture of adherent cells.