Tlačiť
Multipanel test strips for drug testing in urine
Dodávateľ: SURESCREEN
SDA7DS12300 S10DS S7DS16
SURESDA7DS12300EA 326.11 EUR
SURESDA7DS12300 SURES10DS SURES7DS16
Multipanel test strips for drug testing in urine
Testy na drogy
Panels available for testing up to ten drugs. Ideal for pinpointing which drug is being abused. Specials also available depending on quantities, please contact your Avantor office for further information.
  • 99% accurate
  • Clear results in five minutes
  • Wide range of screening options
AMP: amphetamines
BAR: barbiturates
BZO: benzodiazepines
COC: cocaine
mAMP: methamphetamines
MDMA: ecstasy
MTD: methadone
OPI: opiates
PCP: phenylcyclidine
TCA: tricyclic antidepressants
THC: cannabis
BAR: barbiturates
BZO: benzodiazepines
COC: cocaine
mAMP: methamphetamines