Tlačiť
Labelling tapes, temperature resistant
Dodávateľ: PRECISION DYNAMICS TIMEM
AT-312 AT-334 AT-301
817-1581EA 53.35 EUR
817-1581 817-1582 817-1583
Labelling tapes, temperature resistant
Pásky
Textile based with special adhesive. For use in freezing conditions, drying cupboards, incubators, sterilisers, and more.
  • Temperature range −231,5 to +260 °C
  • Can be written on with pencil
Length: 7,6 m
Colour: White