Tlačiť
Tanks, cylindrical, with spigot, lightweight
Dodávateľ: Thermo Fisher Scientific
54102-0005 54102-0007 54102-0010 54102-0015 54102-0030 54102-0055
216-0323EA 364.51 EUR
216-0323 216-0324 216-0325 216-0326 216-0327 216-0328
Tanks, cylindrical, with spigot, lightweight
Zásobníky Zásobníky válcové
Economical tanks with graduations. With protruding rim (flange). Wall thickness ~2,4 mm. LLDPE, translucent with lid and spigot.
  • Good chemical resistance
  • Heat resistant from −100 to +120 °C
  • Conform to the requirements of USP VI and FDA 21 CFR 177.1520