Tlačiť
Disposable syringes, Omnifix®
Dodávateľ: BRAUN
4616308F 4617304F 4616057V 4616103V 4616200V 4617100V 4617029V 4617053V 4617207V 4617509F 4617022V 4616025V 4616502F
612-0115EA 164.86 EUR
612-0115 612-0125 612-2892 612-2893 612-2894 612-2897 612-2895 612-2896 612-2898 612-3355 613-1660 613-3837 612-4871
Disposable syringes, Omnifix®
Stříkačky Střílkačky zdravotnické
Sterile, disposable, three-piece, made from PP, latex- and PVC-free.
  • No needle, with clearly readable graduations
  • Piston is equipped with a double gasket for slower injection and collection, particularly suitable for dispensing smaller quantities
  • Minimal sample loss
DIN EN ISO 10993
Certifikácie: DIN EN ISO 10993