Tlačiť
Syringes for On-Column GC injection
Dodávateľ: HAMILTON BONADUZ
200740 200741 200742 200743 80386 87404 87405 86257
549-1107EA 188.93 EUR
549-1107 549-1108 549-1109 549-1110 HAMI80386 549-1229 HAMI87405 549-1271
Syringes for On-Column GC injection
Stříkačky Stříkačky chromatografické
GC on-column syringes with three choices of needle: Stainless steel fixed (N), stainless steel removable (RN) or fused silica removable (RNFS).