Tlačiť
Modified syringes Microliter™7000 series
Dodávateľ: HAMILTON BONADUZ
80100 86200 86211 86250 86259 88000 88011 88400 88500 88511 80135 88411 86276 80176 86274 CAL86200 80175 CAL88011 CAL80100 CAL86250 CAL86211 CAL88500 CAL88400
549-0904EA 140.6 EUR
549-0904 549-1224 549-1225 549-1226 549-1227 549-1235 549-1236 549-1237 549-1239 549-1240 549-1128 549-1238 549-1273 549-0580 549-1272 HAMICAL86200 548-2287 548-2354 HAMICAL80100 HAMICAL86250 HAMICAL86211 HAMICAL88500 HAMICAL88400
Modified syringes Microliter™7000 series
Stříkačky Stříkačky chromatografické
Suitable for applications such as GC, TLC and micro pipetting. Manufactured according to the principle of dead volume by positive pressure. A tungsten piston is introduced into the syringe as far as the tip of the needle. Needle and piston are calibrated against each other and must be changed at the same time in the event of replacement.
  • Volumetric dosage takes place in the needle
  • Precision ±1% of the nominal volume