Tlačiť
Stoppers, rubber, with reversible edge
Dodávateľ: Saint Gobain Life Sciences
407010-50 407013-50 407025-10 407030-05
217-0183EA 56.08 EUR
217-0183 391-2257 217-0185 217-0186
Stoppers, rubber, with reversible edge
Zátky Zátky kaučukové (gumové)
The white stoppers made of natural rubber guarantee absolute impermeability, even when the edges of the tubes or glass, metal or plastic containers to be sealed are imperfect. They can be punctured and resealed after sampling, even after several uses.
  • Outstanding sealing and protection from the outside atmosphere
  • Suitable for puncturing
  • Economical