Tlačiť
Stoppers, rubber, conical, with holes
Dodávateľ: DEUTSCH NEUMANN
1011110 1011112 1011114 1011117 1011118 1011121 1011123 1011126 1011129 1011131 1012123 1012129 1012114 1012117 1012118 1012121
217-9454EA 2.17 EUR
217-9454 217-9455 217-9456 217-9457 217-9458 217-9459 217-9460 217-9461 217-9462 217-9463 217-9464 217-0828 217-0824 217-0825 217-0826 217-0827
Stoppers, rubber, conical, with holes
Zátky Zátky kaučukové (gumové)
Conical natural grey rubber stoppers with one hole.
  • Resistant to common laboratory chemicals
  • Highly elastic, for a perfect seal and exceptional bonding
  • With pre-drilled holes
  • Withstands temperatures from −25 to +70 °C (+120 °C in humid atmosphere)
  • Hardness (shore durometer A): 40
Withstands temperatures from -25 to +70 °C (+120 °C in moist heat)
Hardness (Shore durometer A): 40
DIN 12871
Certifikácie: DIN 12871