Tlačiť
Caps, Versilic®
Dodávateľ: Saint Gobain Life Sciences
162014-100 162016-50 162010-100 162012-100 162025-50 162030-10
217-0003EA 445.25 EUR
217-0003 217-0004 217-0001 217-0002 217-0109 217-0110
Caps, Versilic®
Uzávěry Víčka na zkumavky
Made of translucent silicone. Ideal for protecting piping/tubing or protruding cylindrical components during transport and all operations that require aperture protection. (against knocks, splashes, paint and more).
  • Outstanding sealing and protection from the outside
  • Can be perforated and autoclaved
  • Resistant to ageing, ozone and repeated sterilisation
The size given corresponds to the Ext. Ø of the vessel to be capped in mm. Resistant to temperatures from −60 to +220 °C (up to 250 °C for short periods).