Tlačiť
Stopcocks, one-way
Dodávateľ: LENZ LABORGLAS
02101201 02101202 02101404 02102910 02161201 02161202 02161804 02162106 02101402 02101804 02102106 02102408 02241402 02161402 02221402 02161404
227-3130EA 28.6 EUR
227-3130 205-2221 227-3135 227-3140 201-1106 201-0494 201-3116 201-1107 227-3133 205-2223 227-3138 227-3139 201-3114 201-3136 LENZ02221402 LENZ02161404
Stopcocks, one-way
Kohoutky
DURAN®, borosilicate glass, with solid glass or PTFE key.
  • Replaceable NS keys
  • Screw thread retaining device
  • Leakage rate compliant with DIN 12540-1
Straight bore.
* with capillary side arms
** with rubber retaining device.
Certifikácie: DIN 12554.