Tlačiť
Valves, PRODURAN®, DURAN®
Dodávateľ: DWK LIFE SCIENCES
286056005 286057001 286055806 286056202 286056605 286056802 286056408 286057104
201-0012EA 72.6 EUR
201-0012 201-0023 201-0011 201-0013 201-0021 201-0022 201-1319 201-1320
Valves, PRODURAN®, DURAN®
Kohoutky
DURAN®, borosilicate glass 3.3, clear, with PTFE spindle.
  • Available straight or angled
  • Temperature resistance from −20 to +150 °C, high chemical resistance
  • Valve spindles are replaceable
These valve cocks are ideal for exact dispensing and therefore ideal for glass equipment. Easy action valve spindle with a safety device that prevents it from being unintentionally pulled out completely. Re-adjustable seal.