Tlačiť
Magnetic stirring discs
Dodávateľ: COWIE
001.709 001.710 001.720 001.730
442-9832EA 3.32 EUR
442-9832 442-9833 442-9834 442-9835
Magnetic stirring discs
Míchadla Míchadla magnetická
PTFE covered, strong Alnico V magnetic core. Smooth surface. Designed for use in tubes.
  • Excellent chemical resistance
  • Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable