Tlačiť
Magnetic stirring bar retrievers
Dodávateľ: COWIE
004.1300 004.1450 004.1350
442-9912EA 13.68 EUR
442-9912 442-9330 442-0158
Magnetic stirring bar retrievers
Míchadla Nástroje na míchání
PP covered, strong Alnico V magnetic core and hanging ring at one end. For the removal of stirrer magnets from vessels of all kinds.
  • Excellent chemical resistance
  • Temperature resistant, suitable for high and low temperature applications