Tlačiť
Magnetic stirring bars, Komet
Dodávateľ: THERMO H + P LABORTECHNIK
50087924 50087930 50087909 50087902
442-9002EA 93.06 EUR
442-9002 442-9003 442-9004 442-9005
Magnetic stirring bars, Komet
Míchadla Míchadla magnetická
Komet magnetic stirring bars made from SmCo (Samarium Cobalt) have a PTFE coating which means they can be sterilised. Particularly recommended for high speed stirring, liquids with a high viscosity and stirring in vessels where there is a large distance between stirrer and stirring bar, such as measuring cyliners.
  • Magnetic power two to three times higher than that of conventional bars
  • Magnetic properties remain for many years