Tlačiť
Distilling bridges
Dodávateľ: LENZ LABORGLAS
05040001 05040002 05040003 05040004
201-0345EA 37.75 EUR
201-0345 201-0346 201-0347 271-3101
Distilling bridges
Komponenty pro destilaci Nástavce
DURAN®, borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Still heads without condenser, with two vertical cones.