Tlačiť
Gamma radiation sterilisation indicator labels
Dodávateľ: CROSSTEX
CPI-R01 CPI-R02 CPI-R03
489-0091EA 348.89 EUR
489-0091 489-0092 489-0093
Gamma radiation sterilisation indicator labels
Indikátory sterilizace
Self-adhesive, non-toxic, chemical process indicators that change from yellow to red upon exposure to gamma or electron beam radiation. Labels are coated with a UV blocking agent for protection from exposure to UV light. A minimum of 15 kGy irradiation is recommended for sufficient end-point colour development. At greater dosage levels, the radiation indicators exhibit a darker shade of red.
  • When exposed, imprinted indicators (Cat. No. 489-0093) read 'RED IS γ-EXPOSED'
  • Stable for at least 24 months from date of manufacture when stored at room temperature
Certifikácie: Conform to AAMI ST60 and ISO 11140-1, Section 6 for Class 1 Process Indicators.