Tlačiť
Spore strips, Sportrol™
Dodávateľ: CROSSTEX
DS-100 STN-062 STS-062 BG-105 BG-106 BS-106 PM-106
489-0077EA 784.2 EUR
489-0077 489-0076 489-0096 NAMSBG-105 NAMSBG-106 NAMSBS-106 NAMSPM-106
Spore strips, Sportrol™
Indikátory sterilizace
Biological sterilisation indicators are used to perform sterilisation cycle validation. They are created by inoculating spore suspensions of certified population and resistance onto paper strips.
  • Standard spore strips and combination spore strips are 7×38 mm
  • Mini spore strips are 2×10 mm in size and will fit into small areas of a device where a standard sized spore strip cannot be used, such as within a syringe barrel, inside tubing or under a cap
  • All spore strips have a shelf life of 24 months when stored under labelled conditions, except Geobacillus stearothermophilus strips 489-0081 and 489-0096 (18 months)
Meet USP and ISO 11128-1 standards.
Certifikácie: Meet USP and ISO 11138-1 standards.
Objednávacie informácie: Spore strips are individually packaged in glassine envelopes in packs of 100. Supplied with instructions for use, certified population, D-value, Z-value, survival/kill data, and expiry date.