Tlačiť
Chemical spoon, double-ended
Dodávateľ: RSG
023.250 023.120 023.210 023.300 023.180 023.150
231-0092EA 20.63 EUR
231-0092 231-1221 231-0189 231-2275 231-2274 231-2273
Chemical spoon, double-ended
Lžíce
Stainless steel 18/10, anti-magnetic. Multipurpose spoon for a wide range of uses. With cylindrical handle.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean