Tlačiť
Cover spectacles, Squale
Dodávateľ: BOLLE
SQUPSI SQUWPCC5
111-0387EA 12.27 EUR
111-0387 BOLESQUWPCC5
Cover spectacles, Squale
Brýle
Cover glasses (OTG) give excellent protection for wearers of prescription eyewear. Supplied with adjustable cord.
  • Adjustable Nylon temples
  • Moulded lens, brow guard and side shields
  • Indirect ventilation
Weight: 43 g
Certifikácie: EN 166, EN 170