Tlačiť
Drigalski spatulas
Dodávateľ: RSG
353.165 354.165 80354195
231-2185EA 14.76 EUR
231-2185 231-1085 231-0652
Drigalski spatulas
Špachtle
Stainless steel 18/10, anti-magnetic.
  • For bacteriological applications
  • Can also be used as stirrer
  • For even spreading or better distribution across media