Tlačiť
Double spatulas, micro
Dodávateľ: RSG
385.130 385.185 385.210 387.250 387.185 387.150 385.150
231-2261EA 32.33 EUR
231-2261 231-2262 231-2263 231-0401 231-0400 231-0399 231-0296
Double spatulas, micro
Špachtle
Stainless steel 18/10, anti-magnetic. Multipurpose spatula for a wide range of uses.
  • Both ends have an angled spoon
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean