Tlačiť
Spoon spatulas
Dodávateľ: MORGAN ADVANCED MATERIALS HALD
L74/2 L74/4 L74/6 L74/8
231-1342EA 23.99 EUR
231-1342 231-1344 231-1346 231-1348
Spoon spatulas
Špachtle
Porcelain, glazed white.
  • Broad shape
  • Excellent chemical resistance
  • Heat resistant up to 1000 °C
  • Designed for rough handling