Tlačiť
Sealing film for PCR plates, Axygen® CyclerSeal
Dodávateľ: Corning
PCR-TS PCR-TS-900
732-7508EA 76.21 EUR
732-7508 732-1381
Sealing film for PCR plates, Axygen® CyclerSeal
Těsnění Těsnění mikrodestičky
Safeguards samples during transport and storage. Ideal for sealing PCR microplates, but also suitable for other microplates.
  • When used with a compression mat, eliminates well-to-well contamination and cross-over
  • Functional temperature range: −40 to +104 °C
  • Colour: transparent