Tlačiť
Sealing film, Axygen® AxySeal
Dodávateľ: Corning
PCR-SP PCR-SP-S
732-7506EA 58.97 EUR
732-7506 732-7507
Sealing film, Axygen® AxySeal
Těsnění Těsnění mikrodestičky
Polyester based with uniformly and consistently applied acrylic adhesive to eliminate the edge effect in ELISA assays. Suitable for sealing tissue culture plates, for short-term storage and incubation, for transport and the containment of biohazardous solutions.
  • No edge effect in ELISA assays
  • Functional temperature range of −40 to +104 °C